Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrphostin AG528 97.0+%, TCI America™
SDP

Catalog No. T350425MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T350425MG 25 mg
T3504100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T350425MG Supplier TCI America Supplier No. T350425MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133550-49-9
Molecular Formula C18H14N2O3
Molecular Weight (g/mol) 306.321
MDL Number MFCD01076474
InChI Key MYKCTDWWIWGLHW-UHFFFAOYSA-N
Synonym (E)-3-(3,4-Dihydroxyphenyl)-2-(indoline-1-carbonyl)acrylonitrile
PubChem CID 4886983
IUPAC Name 2-(2,3-dihydroindole-1-carbonyl)-3-(3,4-dihydroxyphenyl)prop-2-enenitrile
SMILES C1CN(C2=CC=CC=C21)C(=O)C(=CC3=CC(=C(C=C3)O)O)C#N

Specifications

Melting Point 215°C
Quantity 25 mg
Formula Weight 306.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tyrphostin AG528
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.