accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCH3OHNBrCl

CAS RN 13360-45-7

IUPAC Name: 1-(4-bromo-3-chlorophenyl)-3-methoxy-3-methylurea
Synonyms: chlorbromuron
Molecular Weight (g/mol): 293.55
Molecular Formula: C9H10BrClN2O2
InChi Key: NLYNUTMZTCLNOO-UHFFFAOYSA-N
SMILES: CON(C)C(=O)NC1=CC=C(Br)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results