accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCl

CAS RN 13361-36-9

IUPAC Name: 2-methoxyethyl 2-chloroacetate
Synonyms: 2-methoxyethyl 2-chloroacetate
Molecular Weight (g/mol): 152.57
Molecular Formula: C5H9ClO3
InChi Key: SVLBINWLZIVRJX-UHFFFAOYSA-N
SMILES: COCCOC(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results