Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethyl Chloroacetate 97.0+%, TCI America™
SDP

Catalog No. C13465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13465G Supplier TCI America Supplier No. C13465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13361-36-9
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.57
MDL Number MFCD00191626
InChI Key SVLBINWLZIVRJX-UHFFFAOYSA-N
Synonym Chloroacetic Acid 2-Methoxyethyl Ester
PubChem CID 221430
IUPAC Name 2-methoxyethyl 2-chloroacetate
SMILES COCCOC(=O)CCl

Specifications

Color Colorless
Boiling Point 98°C
Quantity 5 g
Formula Weight 152.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Chloroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.