accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN(E)

CAS RN 13362-78-2

IUPAC Name: 4-[(1E)-2-(pyridin-4-yl)ethenyl]pyridine
Synonyms: 4-[(1E)-2-(pyridin-4-yl)ethenyl]pyridine
Molecular Weight (g/mol): 182.23
Molecular Formula: C12H10N2
InChi Key: MGFJDEHFNMWYBD-OWOJBTEDSA-N
SMILES: C(=C/C1=CC=NC=C1)\C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)