Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(4-pyridyl)ethylene, MP Biomedicals

Catalog No. ICN212576 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212576 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212576 Supplier MP Biomedicals, Inc Supplier No. 0521257610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13362-78-2
Molecular Formula C12H10N2
Molecular Weight (g/mol) 182.226
InChI Key MGFJDEHFNMWYBD-OWOJBTEDSA-N
Synonym 4,4'-vinylenedipyridine, 1,2-di 4-pyridyl ethylene, 1,2-bis 4-pyridyl ethylene, 1,2-di pyridin-4-yl ethene, trans-1,2-bis 4-pyridyl ethylene, 1,2-di-4-pyridylethene, e-1,2-di pyridin-4-yl ethene, pyridine, 4,4'-1e-1,2-ethenediylbis, 4-4-pyridylvinyl pyridine, pyridine, 4,4'-1,2-ethenediyl bis-, e
PubChem CID 776222
IUPAC Name 4-[(E)-2-pyridin-4-ylethenyl]pyridine
SMILES C1=CN=CC=C1C=CC2=CC=NC=C2

Specifications

Quantity 10 g
Formula Weight 182.23
Chemical Name or Material 1, 2-Bis(4-pyridyl)ethylene
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.