accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1336893-15-2

IUPAC Name: 5,11-dibromo-2,8-bis[(2-butyloctyl)oxy]-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 5,11-dibromo-2,8-bis[(2-butyloctyl)oxy]-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 716.72
Molecular Formula: C34H52Br2O2S2
InChi Key: XXZPBOPQASGASH-UHFFFAOYNA-N
SMILES: CCCCCCC(CCCC)COC1=C2C=C(Br)SC2=C(OCC(CCCC)CCCCCC)C2=C1SC(Br)=C2
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results