Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromo-4,8-bis[(2-butyl-n-octyl)oxy]benzo[1,2-b:4,5-b']dithiophene 94.0+%, TCI America™
SDP

Catalog No. D4592200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4592200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4592200MG Supplier TCI America Supplier No. D4592200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1336893-15-2
Molecular Formula C34H52Br2O2S2
Molecular Weight (g/mol) 716.716
InChI Key XXZPBOPQASGASH-UHFFFAOYSA-N
PubChem CID 118475389
IUPAC Name 2,6-dibromo-4,8-bis(2-butyloctoxy)thieno[2,3-f][1]benzothiole
SMILES CCCCCCC(CCCC)COC1=C2C=C(SC2=C(C3=C1SC(=C3)Br)OCC(CCCC)CCCCCC)Br

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 716.72
Percent Purity ≥94.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 2,6-Dibromo-4,8-bis[(2-butyl-n-octyl)oxy]benzo[1,2-b:4,5-b′]dithiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.