accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13371-95-4

IUPAC Name: N-benzyl-2-ethoxyaniline
Synonyms: N-benzyl-2-ethoxyaniline
Molecular Weight (g/mol): 227.31
Molecular Formula: C15H17NO
InChi Key: VCFBFZSSLLVLIS-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results