Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-o-phenetidine 98.0+%, TCI America™
SDP

Catalog No. B043125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B043125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B043125G Supplier TCI America Supplier No. B043125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13371-95-4
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.307
MDL Number MFCD00059373
InChI Key VCFBFZSSLLVLIS-UHFFFAOYSA-N
Synonym N-Benzyl-2-ethoxyaniline
PubChem CID 563482
IUPAC Name N-benzyl-2-ethoxyaniline
SMILES CCOC1=CC=CC=C1NCC2=CC=CC=C2

Specifications

Melting Point 32°C
Quantity 25 g
Formula Weight 227.31
Percent Purity ≥98.0% (T)
Physical Form Crystals
Chemical Name or Material N-Benzyl-o-phenetidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.