CAS RN 1337531-89-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PERK Inhibitor I, GSK2606414
GSK2606414 is a cell-permeable, highly potent inhibitor of EIF2AK3/PERK (IC50 = 0.4 nM; [ATP] = 5 μM). Targets PERK in its inactive DFG conformation at the ATP-binding region.