Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PERK Inhibitor I, GSK2606414

Catalog No. 5165355MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-653-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-653-55MG Supplier MilliporeSigma™ Supplier No. 5165355MG
Add to Cart
Add to Cart

GSK2606414 is a cell-permeable, highly potent inhibitor of EIF2AK3/PERK (IC50 = 0.4 nM; [ATP] = 5 μM). Targets PERK in its inactive DFG conformation at the ATP-binding region.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins Perk
CAS 1337531-89-1
Formula Weight 451.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.