CAS RN 13380-84-2
CAS RN 13380-84-2
IUPAC Name:
4-(hydroxymethyl)cyclohexane-1-carboxylic acid
Synonyms:
4-(hydroxymethyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
158.2
Molecular Formula:
C8H14O3
InChi Key:
VQMIUUBKKPIDBN-UHFFFAOYSA-N
SMILES:
OCC1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
4-(Hydroxymethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
CAS: 13380-84-2 Molecular Formula: C8H14O3 Molecular Weight (g/mol): 158.20 MDL Number: MFCD10566907,MFCD01734677,MFCD10566906 InChI Key: VQMIUUBKKPIDBN-UHFFFAOYSA-N Synonym: 4-Carboxy-1-cyclohexanemethanol PubChem CID: 202819 IUPAC Name: 4-(hydroxymethyl)cyclohexane-1-carboxylic acid SMILES: OCC1CCC(CC1)C(O)=O