Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. H10155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10155G 5 g
H101525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10155G Supplier TCI America Supplier No. H10155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13380-84-2
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD10566907,MFCD01734677,MFCD10566906
InChI Key VQMIUUBKKPIDBN-UHFFFAOYSA-N
Synonym 4-Carboxy-1-cyclohexanemethanol
PubChem CID 202819
IUPAC Name 4-(hydroxymethyl)cyclohexane-1-carboxylic acid
SMILES OCC1CCC(CC1)C(O)=O

Specifications

Color Red-White
Quantity 5 g
Formula Weight 158.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hydroxymethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture)
RTECSNumber GU8582500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.