accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2OO

CAS RN 13388-33-5

IUPAC Name: 2-ethenyl-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-ethenylanthracene-9,10-dione
Molecular Weight (g/mol): 234.25
Molecular Formula: C16H10O2
InChi Key: IQOUOAIZDKROQT-UHFFFAOYSA-N
SMILES: C=CC1=CC2=C(C=C1)C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results