accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOH3CNHNOH3CHNO(E/Z)

CAS RN 1338806-73-7

IUPAC Name: 3-[3-(2-{4-[(2,6-dimethylmorpholin-4-yl)methyl]phenyl}ethenyl)-1H-indazol-6-yl]-5'-methoxy-1',2'-dihydrospiro[cyclopropane-1,3'-indol]-2'-one
Synonyms: 3-[3-(2-{4-[(2,6-dimethylmorpholin-4-yl)methyl]phenyl}ethenyl)-1H-indazol-6-yl]-5'-methoxy-1'H-spiro[cyclopropane-1,3'-indol]-2'-one
Molecular Weight (g/mol): 534.66
Molecular Formula: C33H34N4O3
InChi Key: DADASRPKWOGKCU-UHFFFAOYNA-N
SMILES: COC1=CC2=C(NC(=O)C22CC2C2=CC=C3C(NN=C3C=CC3=CC=C(CN4CC(C)OC(C)C4)C=C3)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results