accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1339138-20-3

IUPAC Name: N-(3-amino-2-methylpropyl)acetamide
Synonyms: N-(3-amino-2-methylpropyl)acetamide
Molecular Weight (g/mol): 130.19
Molecular Formula: C6H14N2O
InChi Key: KPYAYJPVNXTYPE-UHFFFAOYNA-N
SMILES: CC(CN)CNC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results