Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-2-methyl-1,3-propanediamine 94.0+%, TCI America™
SDP

Catalog No. A19045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19045G Supplier TCI America Supplier No. A19045G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1339138-20-3
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD08276270
InChI Key KPYAYJPVNXTYPE-UHFFFAOYNA-N
Synonym 1-Acetamido-3-amino-2-methylpropane, N-(3-Amino-2-methylpropyl)acetamide
PubChem CID 44629783
IUPAC Name N-(3-amino-2-methylpropyl)acetamide
SMILES CC(CN)CNC(C)=O

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 130.19
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Acetyl-2-methyl-1,3-propanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.