accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13392-18-2

IUPAC Name: [2-(3,5-dihydroxyphenyl)-2-hydroxyethyl][1-(4-hydroxyphenyl)propan-2-yl]azanium bromide
Synonyms: fenoterol
Molecular Weight (g/mol): 384.27
Molecular Formula: C17H22BrNO4
InChi Key: SGZRQMALQBXAIQ-UHFFFAOYNA-N
SMILES: [Br-].CC(CC1=CC=C(O)C=C1)[NH2+]CC(O)C1=CC(O)=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results