Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenoterol hydrobromide, MP Biomedicals™

Catalog No. ICN15803991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15803991 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15803991 Supplier MP Biomedicals, Inc Supplier No. 0215803991
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13392-18-2
Molecular Formula C17H21NO4
Molecular Weight (g/mol) 303.358
InChI Key LSLYOANBFKQKPT-UHFFFAOYSA-N
Synonym fenoterol, phenoterol, fenoterolum, berotec, fenoterolum inn-latin, berotek, p-hydroxyphenylorciprenaline, p-hydroxyphenyl-orciprenaline, fenoterol usan:ban:inn, 1-3,5-dihydroxyphenyl-1-hydroxy-2-4-hydroxyphenyl isopropylamino ethane
PubChem CID 3343
ChEBI CHEBI:149226
IUPAC Name 5-[1-hydroxy-2-[1-(4-hydroxyphenyl)propan-2-ylamino]ethyl]benzene-1,3-diol
SMILES CC(CC1=CC=C(C=C1)O)NCC(C2=CC(=CC(=C2)O)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 384.3
Physical Form Powder
Chemical Name or Material Fenoterol hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.