accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSSH3CCH3OHO

CAS RN 133944-74-8

IUPAC Name: 2-[(ethoxymethanethioyl)sulfanyl]-2-methylpropanoic acid
Synonyms: 2-[(ethoxymethanethioyl)sulfanyl]-2-methylpropanoic acid
Molecular Weight (g/mol): 208.29
Molecular Formula: C7H12O3S2
InChi Key: FBDXTAKUEUFSGH-UHFFFAOYSA-N
SMILES: CCOC(=S)SC(C)(C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results