accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHCl

CAS RN 13401-56-4

IUPAC Name: 3-chloro-2,2-dimethylpropan-1-ol
Synonyms: 3-chloro-2,2-dimethylpropan-1-ol
Molecular Weight (g/mol): 122.59
Molecular Formula: C5H11ClO
InChi Key: CAZPRAORHCOIHC-UHFFFAOYSA-N
SMILES: CC(C)(CO)CCl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results