accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClOCl

CAS RN 134031-24-6

IUPAC Name: 2,4-dichloropyridine-3-carbaldehyde
Synonyms: 2,4-dichloropyridine-3-carbaldehyde
Molecular Weight (g/mol): 176
Molecular Formula: C6H3Cl2NO
InChi Key: GWBHJHIZHNTJLA-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)C(Cl)=NC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)