CAS RN 1341-36-2
MP Biomedicals, Inc Chromanone, MP Biomedicals
CAS: 1341-36-2 Molecular Formula: C9H8O2 Molecular Weight (g/mol): 148.161 InChI Key: MSTDXOZUKAQDRL-UHFFFAOYSA-N Synonym: 4-chromanone,chroman-4-one,chromanone,4h-1-benzopyran-4-one, 2,3-dihydro,2,3-dihydro-4h-chromen-4-one,2,3-dihydro-1-benzopyran-4-one,isochromanone,3,4-dihydro-2h-1-benzopyran-4-one,2,3-dihydro-4h-1-benzopyran-4-one,dihydrochromone PubChem CID: 68110 IUPAC Name: 2,3-dihydrochromen-4-one SMILES: C1COC2=CC=CC=C2C1=O