Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromanone, MP Biomedicals

Catalog No. ICN224042 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224042 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224042 Supplier MP Biomedicals, Inc Supplier No. 0522404210

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1341-36-2
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
InChI Key MSTDXOZUKAQDRL-UHFFFAOYSA-N
Synonym 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone
PubChem CID 68110
IUPAC Name 2,3-dihydrochromen-4-one
SMILES C1COC2=CC=CC=C2C1=O

Specifications

Quantity 10 g
Formula Weight 146.16
Chemical Name or Material Chromanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.