accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134150-01-9

IUPAC Name: (4-propylphenyl)boronic acid
Synonyms: 4-propylphenylboronic acid
Molecular Weight (g/mol): 164.01
Molecular Formula: C9H13BO2
InChi Key: WLCGYIWOKVWFLB-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)B(O)O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results