Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Propylphenylboronic acid, 95%, Thermo Scientific™

Catalog No. AC393880010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393880010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC393880010 Supplier Thermo Scientific Chemicals Supplier No. 393880010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 134150-01-9
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD00859261
InChI Key WLCGYIWOKVWFLB-UHFFFAOYSA-N
Synonym 4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid
PubChem CID 4100861
IUPAC Name (4-propylphenyl)boronic acid
SMILES CCCC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3(CH2)2C6H4B(OH)2
Quantity 1 g
Formula Weight 164.01
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Propylphenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.