accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHSNHNH2

CAS RN 13431-36-2

IUPAC Name: 3-amino-1-(propan-2-yl)thiourea
Synonyms: 3-amino-1-isopropylthiourea
Molecular Weight (g/mol): 133.21
Molecular Formula: C4H11N3S
InChi Key: WMFXGKCDSJXKHO-UHFFFAOYSA-N
SMILES: CC(C)NC(=S)NN
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results