Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropyl-3-thiosemicarbazide, TCI America™
SDP

Catalog No. I04181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04181G Supplier TCI America Supplier No. I04181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13431-36-2
Molecular Formula C4H11N3S
Molecular Weight (g/mol) 133.213
MDL Number MFCD00025144
InChI Key WMFXGKCDSJXKHO-UHFFFAOYSA-N
PubChem CID 1809376
IUPAC Name 1-amino-3-propan-2-ylthiourea
SMILES CC(C)NC(=S)NN

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 133.21
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropyl-3-thiosemicarbazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.