accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOSiCH3CH3H3COH3C

CAS RN 13433-42-6

IUPAC Name: dimethyl [(trimethylsilyl)methyl]phosphonate
Synonyms: dimethyl (trimethylsilyl)methylphosphonate
Molecular Weight (g/mol): 196.26
Molecular Formula: C6H17O3PSi
InChi Key: QBHFBIHXALNDCB-UHFFFAOYSA-N
SMILES: COP(=O)(C[Si](C)(C)C)OC
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)