Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl trimethylsilylmethylphosphonate, 96%, Thermo Scientific™

Catalog No. AAH2690603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2690603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2690603 Supplier Thermo Scientific Chemicals Supplier No. H2690603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13433-42-6
Molecular Formula C6H17O3PSi
Molecular Weight (g/mol) 196.258
MDL Number MFCD00042921
InChI Key QBHFBIHXALNDCB-UHFFFAOYSA-N
Synonym dimethyl trimethylsilylmethylphosphonate, acmc-20al6f, dimethyl trimethylsilylmethyl phosphonate, dimethoxyphosphorylmethyl trimethyl silane, dimethyl trimethylsilyl methyl phosphonate, dimethyl trimethylsilyl methylphosphonate, trimethylsilylmethylphosphonic acid dimethyl ester, trimethylsilyl methyl-phosphonic acid dimethyl ester, phosphonic acid,p-trimethylsilyl methyl-, dimethyl ester
PubChem CID 4305701
IUPAC Name dimethoxyphosphorylmethyl(trimethyl)silane
SMILES COP(=O)(C[Si](C)(C)C)OC

Specifications

Boiling Point 90°C to 95°C (10 mmHg)
Flash Point 76°C (169°F)
Quantity 1 g
Formula Weight 196.26
Percent Purity 96%
Chemical Name or Material Dimethyl trimethylsilylmethylphosphonate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.