accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 13438-50-1

IUPAC Name: 3-bromofluoranthene
Synonyms: 3-bromofluoranthene
Molecular Weight (g/mol): 281.15
Molecular Formula: C16H9Br
InChi Key: WCXFCLXZMIFHBU-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C3=CC=CC=C3C3=C2C1=CC=C3
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results