Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromofluoranthene 98.0+%, TCI America™
SDP

Catalog No. B45995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45995G 5 g
B45991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45995G Supplier TCI America Supplier No. B45995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13438-50-1
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.15
MDL Number MFCD07787460
InChI Key WCXFCLXZMIFHBU-UHFFFAOYSA-N
PubChem CID 13221800
IUPAC Name 3-bromofluoranthene
SMILES BrC1=CC=C2C3=CC=CC=C3C3=C2C1=CC=C3

Specifications

Melting Point 108°C
Boiling Point 207°C
Quantity 5 g
Formula Weight 281.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromofluoranthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.