accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSiOOONaAlCa

CAS RN 1344-01-0

IUPAC Name: aluminium(3+) calcium sodium silicate
Synonyms: aluminium(3+) calcium sodium orthosilicate
Molecular Weight (g/mol): 182.13
Molecular Formula: AlCaNaO4Si
InChi Key: IDZYHGDLWGVHQM-UHFFFAOYSA-N
SMILES: [Na+].[Al+3].[Ca++].[O-][Si]([O-])([O-])[O-]
Grade 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)