accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COP(?)

CAS RN 134484-36-9

IUPAC Name: {2'-methoxy-[1,1'-binaphthalen]-2-yl}diphenylphosphane
Synonyms: {2'-methoxy-[1,1'-binaphthalen]-2-yl}diphenylphosphane
Molecular Weight (g/mol): 468.54
Molecular Formula: C33H25OP
InChi Key: KRWTWSSMURUMDE-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results