Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98.0+%, TCI America™
SDP

Catalog No. D2775100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2775100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2775100MG Supplier TCI America Supplier No. D2775100MG

Chemical Identifiers

CAS 134484-36-9
Molecular Formula C33H25OP
Molecular Weight (g/mol) 468.536
MDL Number MFCD00269686
InChI Key KRWTWSSMURUMDE-UHFFFAOYSA-N
Synonym (S)-(-)-MOP
PubChem CID 4190002
IUPAC Name [1-(2-methoxynaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
SMILES COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)P(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 100 mg
Formula Weight 468.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-Diphenylphosphino-2′-methoxy-1,1′-binaphthyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.