accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134486-00-3

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxy-3-iodophenyl)propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-hydroxy-3-iodophenyl)propanoic acid
Molecular Weight (g/mol): 529.33
Molecular Formula: C24H20INO5
InChi Key: ZRJAMVZQFHAZAE-NRFANRHFSA-N
SMILES: OC(=O)[C@H](CC1=CC(I)=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)