accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13450-84-5

IUPAC Name: gadolinium(3+) hexahydrate trichloride
Synonyms: gadolinium(3+) hexahydrate trichloride
Molecular Weight (g/mol): 371.69
Molecular Formula: Cl3GdH12O6
InChi Key: PNYPSKHTTCTAMD-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
Molecular Weight (g/mol) 
  • (7)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (7)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (7)
  • (7)
  • (5)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (3)
  • (3)
  • (7)
  • (3)