Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gadolinium(III) chloride hexahydrate

Catalog No. G75325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G75325G 5 g
G753225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G75325G Supplier Sigma Aldrich Supplier No. G75325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gadolinium(III) chloride hexahydrate (GdCl3.6H2O) can be used as a metal precursor in applications such as dual mode MRI contrasting agents. It can also be used as a catalyst in the preparation of one-pot synthesis ofα-amino nitriles via one pot three component reaction.

Specifications

CAS 13450-84-5
Density 2.42 g/mL (at 25°C)
Molecular Formula Cl3Gd -+ 6H2O
Linear Formula GdCl3 -+ 6H2O
Quantity 5 g
MDL Number MFCD00149732
Synonym Gadolinium trichloride hexahydrate
Molecular Weight (g/mol) 371.7
Percent Purity 99% (titration)
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.