accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13453-80-0

IUPAC Name: lithium(1+) dihydrogen phosphate
Synonyms: lithium(1+) dihydrogen phosphate
Molecular Weight (g/mol): 103.93
Molecular Formula: H2LiO4P
InChi Key: SNKMVYBWZDHJHE-UHFFFAOYSA-M
SMILES: [Li+].OP(O)([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)