Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aluminum Monostearate

Catalog No. 062701KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-270-1KG 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-270-1KG Supplier Sigma Aldrich Supplier No. 062701KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used for the packaging of pharmaceuticals, and in the preparation of colors for cosmetics.

Specifications

CAS 13453-80-0
Density 2.5 g/mL at 25°C
Molecular Formula H2LiO4P
Linear Formula LiH2PO4
MDL Number MFCD00016188
Quantity 25 g
Synonym Aluminum dihydroxide stearate; Stearic acid aluminum dihydroxide salt
Molecular Weight (g/mol) 103.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.