CAS RN 13454-70-1
IUPAC Name:
trimolybdenum(6+) dicerium(3+) dodecaoxidandiide
Synonyms:
trimolybdenum(6+) dicerium(3+) dodecaoxidandiide
Molecular Weight (g/mol):
760.07
Molecular Formula:
Ce2Mo3O12
InChi Key:
BPOYBXJKAISYKA-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo+6].[Mo+6].[Mo+6].[Ce+3].[Ce+3]
1
–
1
of
1
results
1
Cerium molybdenum oxide, 99%
CAS: 13454-70-1 Molecular Formula: Ce2Mo3O12 Molecular Weight (g/mol): 760.07 MDL Number: MFCD00798520 InChI Key: BPOYBXJKAISYKA-UHFFFAOYSA-N Synonym: Cerium molybdate