Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium molybdenum oxide, 99%

Catalog No. AA4021022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4021022 100 g
AA4021014 25 g
AA4021006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4021022 Supplier Thermo Scientific Chemicals Supplier No. 04021022
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13454-70-1 | Ce2Mo3O12 | 760.07 g/mol

Cerium molybdenum oxide is used in the aerospace and coating industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-70-1
Molecular Formula Ce2Mo3O12
Molecular Weight (g/mol) 760.07
MDL Number MFCD00798520
InChI Key BPOYBXJKAISYKA-UHFFFAOYSA-N
Synonym Cerium molybdate

Specifications

Melting Point 973°C
Color Brown
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 760.05
Chemical Name or Material Cerium molybdenum oxide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.