CAS RN 1345472-12-9
CAS RN 1345472-12-9
IUPAC Name:
4-[(2-bromo-6-fluorophenyl)methyl]morpholine
Synonyms:
4-[(2-bromo-6-fluorophenyl)methyl]morpholine
Molecular Weight (g/mol):
274.13
Molecular Formula:
C11H13BrFNO
InChi Key:
CWQNAYDNRWPWBM-UHFFFAOYSA-N
SMILES:
FC1=C(CN2CCOCC2)C(Br)=CC=C1
1
–
1
of
1
results
1
4-(2-Bromo-6-fluorobenzyl)morpholine, 96%, Thermo Scientific™
CAS: 1345472-12-9 Molecular Formula: C11H13BrFNO Molecular Weight (g/mol): 274.133 InChI Key: CWQNAYDNRWPWBM-UHFFFAOYSA-N Synonym: 1-bromo-3-fluoro-2-morpholinomethyl benzene,4-2-bromo-6-fluorophenyl methyl morpholine,acmc-209bvy PubChem CID: 56776947 IUPAC Name: 4-[(2-bromo-6-fluorophenyl)methyl]morpholine SMILES: C1COCCN1CC2=C(C=CC=C2Br)F