accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNOPSOH3COH3CNCH3

CAS RN 13457-18-6

IUPAC Name: ethyl 2-{[diethoxy(sulfanylidene)-λ⁵-phosphanyl]oxy}-5-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate
Synonyms: pyrazophos
Molecular Weight (g/mol): 373.36
Molecular Formula: C14H20N3O5PS
InChi Key: JOOMJVFZQRQWKR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2N=C(OP(=S)(OCC)OCC)C=C2N=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results