accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13463-40-6

IUPAC Name: pentakis(methanidylidyneoxidanium) iron
Synonyms: iron pentacarbonyl
Molecular Weight (g/mol): 195.9
Molecular Formula: C5FeO5
InChi Key: FYOFOKCECDGJBF-UHFFFAOYSA-N
SMILES: [Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (4)