Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(0) pentacarbonyl

Catalog No. 48171825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-171-825ML 25 mL
48-171-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-171-825ML Supplier Sigma Aldrich Supplier No. 48171825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron(0) pentacarbonyl

Specifications

CAS 13463-40-6
Melting Point -20°C
Density 1.49 g/mL (at 25°C)
Boiling Point 103°C
Molecular Formula C5FeO5
Refractive Index n20/D 1.5196 (literature)
Linear Formula Fe(CO)5
Quantity 25 mL
MDL Number MFCD00011001
Synonym Iron(0) carbonyl; Pentacarbonyliron(0)
Molecular Weight (g/mol) 195.9
Percent Purity >99.99% trace metals basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.