accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3MgBr

CAS RN 134640-85-0

IUPAC Name: magnesium(2+) 2,3-dimethylbenzen-1-ide bromide
Synonyms: magnesium(2+) 2,3-dimethylbenzen-1-ide bromide
Molecular Weight (g/mol): 209.37
Molecular Formula: C8H9BrMg
InChi Key: PJYPCKZZTIAODG-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC1=C(C)[C-]=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results