CAS RN 13465-58-2
IUPAC Name:
disamarium(3+) trisulfate
Synonyms:
disamarium(3+) trisulfate
Molecular Weight (g/mol):
588.89
Molecular Formula:
O12S3Sm2
InChi Key:
LVSITDBROURTQX-UHFFFAOYSA-H
SMILES:
[Sm+3].[Sm+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
1
–
1
of
1
results
1
Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO)
CAS: 13465-58-2 MDL Number: MFCD00011236