accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOHNN

CAS RN 1346753-05-6

IUPAC Name: 2-(9-oxo-9H-xanthen-2-yl)propanoic acid; 2H,3H,4H,6H,7H,8H,9H,10H-pyrimido[1,2-a]azepine
Synonyms: 1,8-diazabicycloundec-7-ene; 2-(9-oxoxanthen-2-yl)propanoic acid
Molecular Weight (g/mol): 420.51
Molecular Formula: C25H28N2O4
InChi Key: ZLFMLTKPDRGWFB-UHFFFAOYNA-N
SMILES: C1CN=C2CCCCCN2C1.CC(C(O)=O)C1=CC=C2OC3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results